Gene Transcripts

Gene Symbol
Arfrp1
Entrez Gene ID
117051
Gene Name
ADP-ribosylation factor related protein 1
Gene Aliases
Chromosome Location
Chr. 3: 176857667 - 176865105 on Build Rnor_6.0
UniGene ID
Rn.10973
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053980.1 NP_446432.1 3 251
XM_006235718.2 XP_006235780.1 3 247
XM_006235720.2 XP_006235782.1 3 204
GenBank mRNA X78603.1 - 3 251

View Full Product Details