Gene Transcripts
- Gene Symbol
- St8sia2
- Entrez Gene ID
- 117523
- Gene Name
- ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2
- Gene Aliases
- Siat8b
- Chromosome Location
- Chr. 1: 135336374 - 135406699 on Build Rnor_6.0
- UniGene ID
- Rn.44391
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_057156.1 |
NP_476497.1
|
3 |
205 |
|
GenBank mRNA |
L13445.1 |
- |
3 |
205 |
|
View Full Product Details