Gene Transcripts

Gene Symbol
Ppbp
Entrez Gene ID
246358
Gene Name
pro-platelet basic protein (chemokine (C-X-C motif) ligand 7)
Gene Aliases
Cxcl7, Nap-2
Chromosome Location
Chr. 14: 18852433 - 18853237 on Build Rnor_6.0
UniGene ID
Rn.67082
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153721.1 NP_714943.1 2 157
GenBank mRNA AF349115.1 - 2 157

View Full Product Details