Gene Transcripts

Gene Symbol
Cxcl9
Entrez Gene ID
246759
Gene Name
chemokine (C-X-C motif) ligand 9
Gene Aliases
Mig, Scyb9
Chromosome Location
Chr. 14: 17228832 - 17233743 on Build Rnor_6.0
UniGene ID
Rn.7391
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145672.4 NP_663705.1 2 237
GenBank mRNA AF462610.1 - 2 144
AF537208.1 - 2 237
BC087594.1 - 186 7310193

View Full Product Details