Gene Transcripts
- Gene Symbol
- RGD1566134
- Entrez Gene ID
- 298109
- Gene Name
- similar to alpha-2u-globulin
- Gene Aliases
- Chromosome Location
- Chr. 5: 77431489 - 77433910 on Build Rnor_6.0
- UniGene ID
- Rn.86422
- Species
- Rattus norvegicus
- Gene Symbol
- Obp3
- Entrez Gene ID
- 259247
- Gene Name
- alpha-2u globulin PGCL4
- Gene Aliases
- Chromosome Location
- Chr. 5: 77295807 - 77297231 on Build Rnor_6.0
- UniGene ID
- Rn.86413
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB039825.1 |
- |
|
373 |
|
J00738.1 |
- |
|
392 |
|
View Full Product Details