Gene Transcripts

Gene Symbol
Olr1309
Entrez Gene ID
300610
Gene Name
olfactory receptor 1309
Gene Aliases
Chromosome Location
Chr. 8: 43355060 - 43356004 on Build Rnor_6.0
UniGene ID
Rn.143486
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000466.1 NP_001000466.1 1 902

View Full Product Details