Gene Transcripts

Gene Symbol
Wdfy1
Entrez Gene ID
301549
Gene Name
WD repeat and FYVE domain containing 1
Gene Aliases
Chromosome Location
Chr. 9: 85477070 - 85528890 on Build Rnor_6.0
UniGene ID
Rn.8557
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008331.1 NP_001008332.1 6, 7 714
XM_006245190.2 XP_006245252.1 7, 8 798
GenBank mRNA BC085807.1 - 6, 7 714

View Full Product Details