Gene Transcripts

Gene Symbol
HIST1H4G
Entrez Gene ID
8369
Gene Name
histone cluster 1, H4g
Gene Aliases
H4/l, H4FL
Chromosome Location
Chr. 6: 26246611 - 26246977 on Build GRCh38
UniGene ID
Hs.553507
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003547.2 NP_003538.1 1 92
GenBank mRNA BC126276.1 - 1 146
BC126278.1 - 1 146 8991994

View Full Product Details