Gene Transcripts
- Gene Symbol
- Arhgap20
- Entrez Gene ID
- 367085
- Gene Name
- Rho GTPase activating protein 20
- Gene Aliases
- RARhoGAP
- Chromosome Location
- Chr. 8: 56179586 - 56264343 on Build Rnor_6.0
- UniGene ID
- Rn.20805
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_213629.1 |
NP_998794.1
|
15 |
3479 |
|
GenBank mRNA |
AY501475.1 |
- |
15 |
3479 |
|
View Full Product Details