Gene Transcripts

Gene Symbol
Defb14
Entrez Gene ID
641627
Gene Name
defensin beta 14
Gene Aliases
Chromosome Location
Chr. 16: 75647523 - 75650913 on Build Rnor_6.0
UniGene ID
Rn.160439
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037504.1 NP_001032593.1 2 156
XM_006253372.2 XP_006253434.1 3 706
GenBank mRNA AY621346.1 - 2 156

View Full Product Details