Gene Transcripts

Gene Symbol
HBZ
Entrez Gene ID
3050
Gene Name
hemoglobin subunit zeta
Gene Aliases
HBZ-T1, HBZ1
Chromosome Location
Chr. 16: 142422 - 154511 on Build GRCh38
UniGene ID
Hs.585357
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005332.2 NP_005323.1 3 474
XM_005255287.3 XP_005255344.1 4 1104
XM_005255288.3 XP_005255345.1 4 1221
GenBank mRNA BC027892.1 - 3 454
M24173.1 - 3 448

View Full Product Details