Gene Transcripts
- Gene Symbol
- Igkv6-14
- Entrez Gene ID
- 667881
- Gene Name
- immunoglobulin kappa variable 6-14
- Gene Aliases
- Igk-V19-14
- Chromosome Location
- Chr. 6: 70434952 - 70435476 on Build GRCm38
- UniGene ID
- Mm.426557
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U59155.1 |
- |
|
148 |
|
- Gene Symbol
- Igkv6-15
- Entrez Gene ID
- 108022
- Gene Name
- immunoglobulin kappa variable 6-15
- Gene Aliases
- Igk-V19-15
- Chromosome Location
- Chr. 6: 70406469 - 70406992 on Build GRCm38
- UniGene ID
- Mm.426557
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U55641.1 |
- |
1 |
138 |
|
View Full Product Details