Gene Transcripts

Gene Symbol
Igfbp5
Entrez Gene ID
25285
Gene Name
insulin-like growth factor binding protein 5
Gene Aliases
IGF-BP5
Chromosome Location
Chr. 9: 80154230 - 80166813 on Build Rnor_6.0
UniGene ID
Rn.1593
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012817.1 NP_036949.1 2 1100
GenBank mRNA BC087030.1 - 1302
M62781.1 - 2 1100

View Full Product Details