Gene Transcripts

Gene Symbol
Ptpmt1
Entrez Gene ID
29390
Gene Name
protein tyrosine phosphatase, mitochondrial 1
Gene Aliases
Plip
Chromosome Location
Chr. 3: 79734961 - 79743737 on Build Rnor_6.0
UniGene ID
Rn.108023
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105726.1 NP_001099196.1 3 945
GenBank mRNA FQ219112.1 - 3 585

View Full Product Details