Gene Transcripts

Gene Symbol
Vipr2
Entrez Gene ID
29555
Gene Name
vasoactive intestinal peptide receptor 2
Gene Aliases
Chromosome Location
Chr. 6: 143937961 - 144007028 on Build Rnor_6.0
UniGene ID
Rn.10011
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017238.1 NP_058934.1 3, 4 369
GenBank mRNA U09631.1 - 3, 4 369
Z25885.1 - 3, 4 322

View Full Product Details