Gene Transcripts

Gene Symbol
IFNA4
Entrez Gene ID
3441
Gene Name
interferon alpha 4
Gene Aliases
IFN-alpha4a, INFA4
Chromosome Location
Chr. 9: 21186618 - 21187599 on Build GRCh38
UniGene ID
Hs.1510
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021068.2 NP_066546.1 1 407
GenBank mRNA BC074965.2 - 1 359
BC074966.2 - 1 359 30915288
BC113640.1 - 1 395 8322692
BC113642.1 - 1 395 8322694
M27318.1 - 1 479

Gene Symbol
IFNA10
Entrez Gene ID
3446
Gene Name
interferon alpha 10
Gene Aliases
IFN-alphaC
Chromosome Location
Chr. 9: 21206181 - 21207143 on Build GRCh38
UniGene ID
Hs.282275
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002171.2 NP_002162.1 1 385
GenBank mRNA BC069409.1 - 1 385
BC103972.2 - 1 385 40015081
BC103973.1 - 1 385 40015082
V00551.1 - 1 385

Gene Symbol
IFNA17
Entrez Gene ID
3451
Gene Name
interferon alpha 17
Gene Aliases
IFN-alphaI, IFNA, INFA, LEIF2C1
Chromosome Location
Chr. 9: 21227243 - 21228222 on Build GRCh38
UniGene ID
Hs.282276
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021268.2 NP_067091.1 1 388
GenBank mRNA BC096732.1 - 1 381
BC098355.1 - 1 381 40015056
M11026.1 - 1 388
M38289.1 - 1 481
M71246.1 - 1 300

Gene Symbol
IFNA7
Entrez Gene ID
3444
Gene Name
interferon alpha 7
Gene Aliases
IFN-alphaJ, IFNA-J
Chromosome Location
Chr. 9: 21201469 - 21202205 on Build GRCh38
UniGene ID
Hs.282274
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021057.2 NP_066401.2 1 379
GenBank mRNA BC074991.2 - 1 377
BC074992.2 - 1 377 30915307
BC113994.1 - 1 379 40084821
BC114000.1 - 1 379 40084820
M34913.1 - 1 379

View Full Product Details