Gene Transcripts
- Gene Symbol
- Hist1h3a
- Entrez Gene ID
- 360198
- Gene Name
- histone cluster 1, H3a
- Gene Aliases
- Chromosome Location
- Chr. 13: 23761885 - 23762386 on Build GRCm38
- UniGene ID
- Mm.221301
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK018952.1 |
- |
1 |
463 |
|
View Full Product Details