Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001300728.1 | NP_001287657.1 | 4 | 1456 | |
NM_015251.2 | NP_056066.2 | 4 | 1516 | ||
GenBank mRNA | AB007891.1 | - | 3 | 1993 | |
AK290943.1 | - | 4 | 1301 | ||
AK301781.1 | - | 3 | 1256 | ||
BC002701.2 | - | 4 | 1333 | ||
BI259534.1 | - | 1 | 457 | ||
CR749457.1 | - | 4 | 1409 |