Gene Transcripts

Gene Symbol
ZNF681
Entrez Gene ID
148213
Gene Name
zinc finger protein 681
Gene Aliases
Chromosome Location
Chr. 19: 23739195 - 23758891 on Build GRCh38
UniGene ID
Hs.399952
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138286.2 NP_612143.2 4 1690
GenBank mRNA AK056088.1 - 3 1488
AK122869.1 - 4 1694
BC136346.1 - 4 1569 9020333

View Full Product Details