Gene Transcripts

Gene Symbol
ZNRF2
Entrez Gene ID
223082
Gene Name
zinc and ring finger 2, E3 ubiquitin protein ligase
Gene Aliases
RNF202
Chromosome Location
Chr. 7: 30284307 - 30367692 on Build GRCh38
UniGene ID
Hs.263912
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147128.3 NP_667339.1 3 1699
GenBank mRNA AF513707.1 - 3 1699
AF527533.1 - 3 1026
AK025240.1 - 2 141
AY054122.1 - 4 463

View Full Product Details