Gene Transcripts
- Gene Symbol
- ROPN1L
- Entrez Gene ID
- 83853
- Gene Name
- rhophilin associated tail protein 1 like
- Gene Aliases
- ASP, RSPH11
- Chromosome Location
- Chr. 5: 10441862 - 10482807 on Build GRCh38
- UniGene ID
- Hs.381089
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK026656.1 |
- |
1 |
1121 |
|
- Gene Symbol
- LOC105369214
- Entrez Gene ID
- 105369214
- Gene Name
- uncharacterized LOC105369214
- Gene Aliases
- Chromosome Location
- Chr. 5: 10462865 - 10464841 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details