Gene Transcripts
- Gene Symbol
- LOC101929154
- Entrez Gene ID
- 101929154
- Gene Name
- uncharacterized LOC101929154
- Gene Aliases
- Chromosome Location
- Chr. 5: 77884656 - 77959096 on Build GRCh38
- UniGene ID
- Hs.252895
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_105012.1 |
|
2 |
276 |
|
NR_105013.1 |
|
2 |
169 |
|
View Full Product Details