Gene Transcripts

Gene Symbol
Gsg2
Entrez Gene ID
14841
Gene Name
germ cell-specific gene 2
Gene Aliases
Haspin
Chromosome Location
Chr. 11: 73135484 - 73138294 on Build GRCm38
UniGene ID
Mm.42045
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010353.2 NP_034483.1 1 1361
GenBank mRNA BC113148.1 - 1 1355
D87326.1 - 1 1361

View Full Product Details