Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005583.4 | NP_005574.2 | 2, 3 | 989 | |
XM_005259912.3 | XP_005259969.1 | 2, 3 | 827 | ||
XM_017026816.1 | XP_016882305.1 | 3, 4 | 1689 | ||
XM_017026817.1 | XP_016882306.1 | 3, 4 | 1542 | ||
GenBank mRNA | BC002796.2 | - | 2, 3 | 696 | |
M22637.1 | - | 2, 3 | 554 |