Gene Transcripts

Gene Symbol
Crtc3
Entrez Gene ID
70461
Gene Name
CREB regulated transcription coactivator 3
Gene Aliases
2610312F20Rik, 6332415K15Rik, AI429792, BC011210, TORC3
Chromosome Location
Chr. 7: 80586627 - 80688942 on Build GRCm38
UniGene ID
Mm.392175
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173863.2 NP_776288.2 13 1641
GenBank mRNA AK144840.1 - 13 1643
BC011210.1 - 13 1480 4195538

View Full Product Details