Gene Transcripts
- Gene Symbol
- SOX7
- Entrez Gene ID
- 83595
- Gene Name
- SRY-box 7
- Gene Aliases
- Chromosome Location
- Chr. 8: 10723768 - 10730574 on Build GRCh38
- UniGene ID
- Hs.709543
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031439.3 |
NP_113627.1
|
2 |
2480 |
|
View Full Product Details