Gene Transcripts
- Gene Symbol
- ROBO1
- Entrez Gene ID
- 6091
- Gene Name
- roundabout guidance receptor 1
- Gene Aliases
- DUTT1, SAX3
- Chromosome Location
- Chr. 3: 78597238 - 79767998 on Build GRCh38
- UniGene ID
- Hs.744218
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002941.3 |
NP_002932.1
|
22, 23 |
3253 |
|
NM_133631.3 |
NP_598334.2
|
20, 21 |
3908 |
|
XM_006713277.2 |
XP_006713340.1
|
21, 22 |
3904 |
|
XM_011533976.1 |
XP_011532278.1
|
23, 24 |
3351 |
|
XM_011533977.2 |
XP_011532279.1
|
23, 24 |
3204 |
|
XM_011533978.1 |
XP_011532280.1
|
22, 23 |
3324 |
|
XM_017006982.1 |
XP_016862471.1
|
22, 23 |
3263 |
|
XM_017006983.1 |
XP_016862472.1
|
20, 21 |
3895 |
|
XM_017006985.1 |
XP_016862474.1
|
21, 22 |
3004 |
|
GenBank mRNA |
AF040990.1 |
- |
21, 22 |
3045 |
|
AK299920.1 |
- |
12, 13 |
1942 |
|
BC112336.1 |
- |
20, 21 |
2961 |
40037251 |
BC115020.1 |
- |
20, 21 |
3009 |
7961997 |
BC157861.1 |
- |
20, 21 |
3205 |
9057080 |
BC171855.1 |
- |
20, 21 |
3205 |
9054509 |
BX538319.1 |
- |
14, 15 |
1964 |
|
View Full Product Details