Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001319051.1 | NP_001305980.1 | 9 | 2487 | |
NM_001319052.1 | NP_001305981.1 | 4 | 391 | ||
NM_054110.4 | NP_473451.3 | 9 | 2132 | ||
XM_005264852.4 | XP_005264909.1 | 9 | 1664 | ||
XM_017005682.1 | XP_016861171.1 | 9 | 9545 | ||
XM_017005683.1 | XP_016861172.1 | 9 | 1695 | ||
XM_017005684.1 | XP_016861173.1 | 9 | 1695 | ||
GenBank mRNA | AB078149.1 | - | 9 | 1657 | |
AJ505964.1 | - | 3 | 265 | ||
AK094780.1 | - | 12 | 1705 | ||
AK312425.1 | - | 9 | 2133 | ||
AL831925.1 | - | 9 | 2160 | ||
AL832575.2 | - | 9 | 1634 | ||
AL832736.1 | - | 9 | 2102 | ||
AY035399.1 | - | 9 | 1940 | ||
AY358443.1 | - | 9 | 2118 | ||
BC014789.1 | - | 9 | 2082 | 4212702 | |
BC030625.1 | - | 4 | 396 |