Gene Transcripts

Gene Symbol
ZNF491
Entrez Gene ID
126069
Gene Name
zinc finger protein 491
Gene Aliases
Chromosome Location
Chr. 19: 11798509 - 11809622 on Build GRCh38
UniGene ID
Hs.631634
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005259730.3 XP_005259787.1 2 3286
GenBank mRNA AK096593.1 - 4 2536

View Full Product Details