Gene Transcripts
- Gene Symbol
- HUNK
- Entrez Gene ID
- 30811
- Gene Name
- hormonally up-regulated Neu-associated kinase
- Gene Aliases
- Chromosome Location
- Chr. 21: 31873315 - 32004064 on Build GRCh38
- UniGene ID
- Hs.109437
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014586.1 |
NP_055401.1
|
11 |
5397 |
|
GenBank mRNA |
AJ271722.1 |
- |
11 |
5397 |
|
View Full Product Details