Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000405.4 | NP_000396.2 | 4 | 940 | |
NM_001167607.1 | NP_001161079.1 | 4 | 730 | ||
GenBank mRNA | AK225978.1 | - | 4 | 868 | |
AK298867.1 | - | 4 | 597 | ||
BC009273.2 | - | 4 | 804 | 4053681 | |
L01439.1 | - | 4 | 865 | ||
M76477.1 | - | 4 | 865 | ||
X16087.1 | - | 4 | 733 | ||
X61095.1 | - | 4 | 796 | ||
X62078.1 | - | 4 | 833 |