Gene Transcripts

Gene Symbol
Ptger1
Entrez Gene ID
19216
Gene Name
prostaglandin E receptor 1 (subtype EP1)
Gene Aliases
42kDa, EP1, Ptgerep1
Chromosome Location
Chr. 8: 83666530 - 83670103 on Build GRCm38
UniGene ID
Mm.213000
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013641.2 NP_038669.1 2 1178
GenBank mRNA AK053616.1 - 952
AK164746.1 - 986
BC138739.1 - 2 1140 8861761
BC138744.1 - 2 1140 8861766
D16338.1 - 2 955

View Full Product Details