Gene Transcripts

Gene Symbol
Dpp9
Entrez Gene ID
224897
Gene Name
dipeptidylpeptidase 9
Gene Aliases
6430584G11Rik, A330078I11, DPP IX, DPRP2
Chromosome Location
Chr. 17: 56186674 - 56218889 on Build GRCm38
UniGene ID
Mm.205535
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172624.3 NP_766212.2 9, 10 1219
GenBank mRNA AK039652.1 - 8, 9 1312
AK050021.1 - 9, 10 1161
AK050023.1 - 9, 10 1221
AK078301.1 - 9, 10 1167
AK131178.1 - 6, 7 3082
BC057631.1 - 9, 10 1218 4191886

View Full Product Details