Gene Transcripts

Gene Symbol
Slc26a10
Entrez Gene ID
216441
Gene Name
solute carrier family 26, member 10
Gene Aliases
C78409, D630010I07
Chromosome Location
Chr. 10: 127172313 - 127180709 on Build GRCm38
UniGene ID
Mm.248373
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177615.3 NP_808283.2 1 151
GenBank mRNA AK052641.1 - 89
BC089587.1 - 1 88 6757541

View Full Product Details