Gene Transcripts

Gene Symbol
5031410I06Rik
Entrez Gene ID
381622
Gene Name
RIKEN cDNA 5031410I06 gene
Gene Aliases
Chromosome Location
Chr. 5: 26098668 - 26107407 on Build GRCm38
UniGene ID
Mm.226878
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207657.3 NP_997540.2 5 2028
GenBank mRNA AK077239.1 - 2030

View Full Product Details