Gene Transcripts
- Gene Symbol
- Gm9801
- Entrez Gene ID
- 330552
- Gene Name
- predicted gene 9801
- Gene Aliases
- A230097C02, EG330552, ENSMUSG00000045813
- Chromosome Location
- Chr. 7: 62192028 - 62278831 on Build GRCm38
- UniGene ID
- Mm.242875
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK086712.1 |
- |
|
2344 |
|
View Full Product Details