Gene Transcripts

Gene Symbol
Slc2a1
Entrez Gene ID
24778
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 1
Gene Aliases
GLUTB, GTG1, Glut1, Gtg3, RATGTG1
Chromosome Location
Chr. 5: 138154677 - 138182897 on Build Rnor_6.0
UniGene ID
Rn.3205
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138827.1 NP_620182.1 7 1100
GenBank mRNA BC061873.1 - 7 1028
M13979.1 - 7 1100

View Full Product Details