Gene Transcripts
- Gene Symbol
- KIR2DL1
- Entrez Gene ID
- 3802
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 1
- Gene Aliases
- CD158A, KIR-K64, KIR221, KIR2DS1, NKAT, NKAT-1, NKAT1, p58.1
- Chromosome Location
- Chr. 19: 54769208 - 54784326 on Build GRCh38
- UniGene ID
- Hs.654605
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014218.2 |
NP_055033.2
|
2, 3 |
115 |
|
XM_011526939.2 |
XP_011525241.1
|
2, 3 |
720 |
|
XM_017026783.1 |
XP_016882272.1
|
2, 3 |
720 |
|
GenBank mRNA |
AF022045.1 |
- |
2, 3 |
84 |
|
AF135560.1 |
- |
|
14 |
|
AF285431.1 |
- |
2, 3 |
110 |
|
AF285432.1 |
- |
1, 2 |
37 |
|
AY366237.1 |
- |
2, 3 |
113 |
|
AY366238.1 |
- |
2, 3 |
59 |
|
AY366239.1 |
- |
2, 3 |
107 |
|
AY366240.1 |
- |
2, 3 |
115 |
|
AY366241.1 |
- |
2, 3 |
114 |
|
AY388471.1 |
- |
2, 3 |
107 |
|
AY789055.1 |
- |
2, 3 |
110 |
|
AY789056.1 |
- |
2, 3 |
77 |
|
BC069344.1 |
- |
2, 3 |
110 |
7262296 |
KJ699235.1 |
- |
2, 3 |
350 |
|
L41267.1 |
- |
2, 3 |
109 |
|
U24076.1 |
- |
2, 3 |
110 |
|
U24078.1 |
- |
2, 3 |
117 |
|
- Gene Symbol
- KIR2DL2
- Entrez Gene ID
- 3803
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 2
- Gene Aliases
- CD158B1, CD158b, NKAT-6, NKAT6, p58.2
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.743949
- Species
- Homo sapiens
- Gene Symbol
- KIR2DL3
- Entrez Gene ID
- 3804
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 3
- Gene Aliases
- CD158B2, CD158b, GL183, KIR-023GB, KIR-K7b, KIR-K7c, KIR2DS3, KIR2DS5, KIRCL23, NKAT, NKAT2, NKAT2A, NKAT2B, p58
- Chromosome Location
- Chr. 19: 54738509 - 54753052 on Build GRCh38
- UniGene ID
- Hs.654608
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015868.2 |
NP_056952.2
|
2, 3 |
114 |
|
GenBank mRNA |
AF022048.1 |
- |
2, 3 |
77 |
|
AF135556.1 |
- |
|
14 |
|
AF135566.1 |
- |
|
14 |
|
AF135567.1 |
- |
|
14 |
|
AF285435.1 |
- |
1, 2 |
37 |
|
AY789057.1 |
- |
2, 3 |
77 |
|
BC032422.1 |
- |
2, 3 |
90 |
5220339 |
BC050730.1 |
- |
2, 3 |
108 |
5219336 |
BC110613.1 |
- |
2, 3 |
91 |
40031619 |
FN395076.1 |
- |
2, 3 |
77 |
|
FN552550.1 |
- |
2, 3 |
59 |
|
L41268.1 |
- |
2, 3 |
114 |
|
L76662.1 |
- |
2, 3 |
88 |
|
L76663.1 |
- |
2, 3 |
88 |
|
U24074.1 |
- |
2, 3 |
98 |
|
U50546.1 |
- |
2, 3 |
88 |
|
U73395.1 |
- |
2, 3 |
98 |
|
- Gene Symbol
- KIR2DS1
- Entrez Gene ID
- 3806
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 1
- Gene Aliases
- CD158H, CD158a, KIR2DP1DL1, KIR2DS4, p50.1
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.744319
- Species
- Homo sapiens
- Gene Symbol
- KIR2DS2
- Entrez Gene ID
- 100132285
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 2
- Gene Aliases
- 183ActI, CD158J, CD158b, KIR-2DS2, NKAT-5, NKAT5, cl-49
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.512572
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001291695.1 |
NP_001278624.1
|
2, 3 |
112 |
|
NM_001291701.1 |
NP_001278630.1
|
2, 3 |
112 |
|
NM_012312.4 |
NP_036444.1
|
2, 3 |
112 |
|
GenBank mRNA |
AF135565.1 |
- |
|
14 |
|
AF285438.1 |
- |
1, 2 |
37 |
|
AJ002102.1 |
- |
2, 3 |
77 |
|
AJ002103.1 |
- |
2, 3 |
77 |
|
AJ002104.1 |
- |
2, 3 |
77 |
|
AJ002105.1 |
- |
2, 3 |
77 |
|
AM409094.1 |
- |
2, 3 |
78 |
|
AY366243.1 |
- |
2, 3 |
87 |
|
AY366244.1 |
- |
2, 3 |
76 |
|
BC108917.1 |
- |
2, 3 |
79 |
40017299 |
BC119631.1 |
- |
2, 3 |
96 |
40115568 |
BC119632.1 |
- |
2, 3 |
96 |
40115570 |
BC125146.1 |
- |
1, 2 |
61 |
|
BC126856.1 |
- |
2, 3 |
79 |
40017301 |
FR750994.1 |
- |
2, 3 |
112 |
|
L41347.1 |
- |
2, 3 |
96 |
|
L76667.1 |
- |
2, 3 |
88 |
|
U24079.1 |
- |
2, 3 |
75 |
|
X89893.1 |
- |
2, 3 |
77 |
|
- Gene Symbol
- KIR2DS4
- Entrez Gene ID
- 3809
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 4
- Gene Aliases
- CD158I, KIR-2DS4, KIR1D, KIR2DS1, KIR412, KKA3, NKAT-8, NKAT8
- Chromosome Location
- Chr. 19: 54832676 - 54848569 on Build GRCh38
- UniGene ID
- Hs.654608
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001281971.1 |
NP_001268900.1
|
2, 3 |
166 |
|
NM_001281972.1 |
NP_001268901.1
|
2, 3 |
166 |
|
NM_012314.5 |
NP_036446.3
|
2, 3 |
166 |
|
GenBank mRNA |
AF002255.1 |
- |
2, 3 |
123 |
|
AF135563.1 |
- |
|
14 |
|
AJ417554.1 |
- |
1, 2 |
57 |
|
AJ417555.1 |
- |
1, 2 |
57 |
|
AJ620615.2 |
- |
1, 2 |
50 |
|
AJ620616.2 |
- |
1, 2 |
50 |
|
AY102623.1 |
- |
2, 3 |
77 |
|
AY366245.1 |
- |
2, 3 |
111 |
|
AY366246.1 |
- |
2, 3 |
84 |
|
AY388472.1 |
- |
2, 3 |
115 |
|
AY789064.2 |
- |
2, 3 |
61 |
|
BC096694.1 |
- |
2, 3 |
88 |
40026979 |
BC101977.1 |
- |
2, 3 |
88 |
40004312 |
BC103693.1 |
- |
2, 3 |
88 |
40004314 |
BC108918.1 |
- |
2, 3 |
79 |
40017300 |
BC125147.1 |
- |
1, 2 |
61 |
|
BC127084.1 |
- |
2, 3 |
94 |
40120468 |
BC144566.1 |
- |
2, 3 |
166 |
9053094 |
BC171788.1 |
- |
2, 3 |
166 |
9054442 |
L76671.1 |
- |
2, 3 |
88 |
|
U24077.1 |
- |
2, 3 |
68 |
|
X94609.1 |
- |
2, 3 |
132 |
|
View Full Product Details