Gene Transcripts
- Gene Symbol
- KIR2DS2
- Entrez Gene ID
- 100132285
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 2
- Gene Aliases
- 183ActI, CD158J, CD158b, KIR-2DS2, NKAT-5, NKAT5, cl-49
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.512572
- Species
- Homo sapiens
- Gene Symbol
- KIR2DL2
- Entrez Gene ID
- 3803
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 2
- Gene Aliases
- CD158B1, CD158b, NKAT-6, NKAT6, p58.2
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.743949
- Species
- Homo sapiens
- Gene Symbol
- KIR2DL3
- Entrez Gene ID
- 3804
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 3
- Gene Aliases
- CD158B2, CD158b, GL183, KIR-023GB, KIR-K7b, KIR-K7c, KIR2DS3, KIR2DS5, KIRCL23, NKAT, NKAT2, NKAT2A, NKAT2B, p58
- Chromosome Location
- Chr. 19: 54738509 - 54753052 on Build GRCh38
- UniGene ID
- Hs.654608
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF022048.1 |
- |
3 |
169 |
|
FN552550.1 |
- |
3 |
151 |
|
U73395.1 |
- |
3 |
190 |
|
- Gene Symbol
- KIR2DS4
- Entrez Gene ID
- 3809
- Gene Name
- killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 4
- Gene Aliases
- CD158I, KIR-2DS4, KIR1D, KIR2DS1, KIR412, KKA3, NKAT-8, NKAT8
- Chromosome Location
- Chr. 19: 54832676 - 54848569 on Build GRCh38
- UniGene ID
- Hs.654608
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF135563.1 |
- |
1 |
106 |
|
View Full Product Details