Gene Transcripts
- Gene Symbol
- HLA-DRB1
- Entrez Gene ID
- 3123
- Gene Name
- major histocompatibility complex, class II, DR beta 1
- Gene Aliases
- DRB1, DRw10, HLA-DR1B, HLA-DRB, SS1
- Chromosome Location
- Chr. 6: 32578769 - 32589836 on Build GRCh38
- UniGene ID
- Hs.534322
- Species
- Homo sapiens
- Gene Symbol
- HLA-DQB1
- Entrez Gene ID
- 3119
- Gene Name
- major histocompatibility complex, class II, DQ beta 1
- Gene Aliases
- CELIAC1, HLA-DQB, HLA-DRB1, IDDM1
- Chromosome Location
- Chr. 6: 32659464 - 32666689 on Build GRCh38
- UniGene ID
- Hs.409934
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M20505.1 |
- |
4 |
660 |
|
- Gene Symbol
- HLA-DRB3
- Entrez Gene ID
- 3125
- Gene Name
- major histocompatibility complex, class II, DR beta 3
- Gene Aliases
- DRB1, DRw10, HLA-DR1B, HLA-DR3B, HLA-DRB, SS1
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.696211
- Species
- Homo sapiens
- Gene Symbol
- HLA-DRB4
- Entrez Gene ID
- 3126
- Gene Name
- major histocompatibility complex, class II, DR beta 4
- Gene Aliases
- DR-4, DR4, DRB4, HLA-DR4B
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.534322
- Species
- Homo sapiens
- Gene Symbol
- HLA-DRB5
- Entrez Gene ID
- 3127
- Gene Name
- major histocompatibility complex, class II, DR beta 5
- Gene Aliases
- HLA-DRB
- Chromosome Location
- Chr. 6: 32517343 - 32530316 on Build GRCh38
- UniGene ID
- Hs.485130
- Species
- Homo sapiens
- Gene Symbol
- HLA-DRB6
- Entrez Gene ID
- 3128
- Gene Name
- major histocompatibility complex, class II, DR beta 6 (pseudogene)
- Gene Aliases
- Chromosome Location
- Chr. 6: 32552713 - 32560002 on Build GRCh38
- UniGene ID
- Hs.723344
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001298.1 |
|
4 |
771 |
|
GenBank mRNA |
AF005487.1 |
- |
4 |
771 |
|
L76565.1 |
- |
4 |
842 |
|
- Gene Symbol
- LOC105369230
- Entrez Gene ID
- 105369230
- Gene Name
- HLA class II histocompatibility antigen, DRB1-7 beta chain
- Gene Aliases
- HLA-DRB1
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.696211
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
HG974601.1 |
- |
4 |
683 |
|
HM067848.1 |
- |
4 |
683 |
|
View Full Product Details