Gene Transcripts

Gene Symbol
HFE
Entrez Gene ID
3077
Gene Name
hemochromatosis
Gene Aliases
HFE1, HH, HLA-H, MVCD7, TFQTL2
Chromosome Location
Chr. 6: 26087281 - 26096216 on Build GRCh38
UniGene ID
Hs.233325
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF115264.1 - 4 443
AF144240.1 - 3 645
AJ404378.1 - 2 321
U60319.1 - 7 2436

View Full Product Details