Gene Transcripts
- Gene Symbol
- CCL3L1
- Entrez Gene ID
- 6349
- Gene Name
- C-C motif chemokine ligand 3 like 1
- Gene Aliases
- 464.2, D17S1718, G0S19-2, LD78, LD78-beta(1-70), LD78BETA, MIP1AP, SCYA3L, SCYA3L1
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.512683
- Species
- Homo sapiens
- Gene Symbol
- CCL3
- Entrez Gene ID
- 6348
- Gene Name
- C-C motif chemokine ligand 3
- Gene Aliases
- G0S19-1, LD78ALPHA, MIP-1-alpha, MIP1A, SCYA3
- Chromosome Location
- Chr. 17: 36088256 - 36090160 on Build GRCh38
- UniGene ID
- Hs.514107
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
D63785.1 |
- |
2, 3 |
168 |
|
- Gene Symbol
- CCL3L3
- Entrez Gene ID
- 414062
- Gene Name
- C-C motif chemokine ligand 3 like 3
- Gene Aliases
- 464.2, D17S1718, G0S19-2, LD78, LD78BETA, SCYA3L, SCYA3L1
- Chromosome Location
- Chr. 17: 36194869 - 36196748 on Build GRCh38
- UniGene ID
- Hs.454713
- Species
- Homo sapiens
View Full Product Details