Gene Transcripts

Gene Symbol
JAK3
Entrez Gene ID
3718
Gene Name
Janus kinase 3
Gene Aliases
JAK-3, JAK3_HUMAN, JAKL, L-JAK, LJAK
Chromosome Location
Chr. 19: 17824782 - 17848071 on Build GRCh38
UniGene ID
Hs.515247
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000215.3 NP_000206.2 23, 24 3315
XM_005259896.3 XP_005259953.1 23, 24 3354
GenBank mRNA BM561964.1 - 1 23
U09607.1 - 22, 23 3310

View Full Product Details