Gene Transcripts
- Gene Symbol
- IL23A
- Entrez Gene ID
- 51561
- Gene Name
- interleukin 23 subunit alpha
- Gene Aliases
- IL-23, IL-23A, IL23P19, P19, SGRF
- Chromosome Location
- Chr. 12: 56334159 - 56340410 on Build GRCh38
- UniGene ID
- Hs.382212
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016584.2 |
NP_057668.1
|
2, 3 |
422 |
|
XM_011538477.2 |
XP_011536779.1
|
4, 5 |
609 |
|
GenBank mRNA |
AB030000.1 |
- |
2, 3 |
399 |
|
AF301620.1 |
- |
2, 3 |
406 |
|
AY359083.1 |
- |
2, 3 |
426 |
|
BC066267.1 |
- |
2, 3 |
289 |
6971848 |
BC066268.1 |
- |
2, 3 |
289 |
6971849 |
BC066269.1 |
- |
2, 3 |
289 |
6971851 |
BC067511.1 |
- |
2, 3 |
289 |
6971850 |
BC067512.1 |
- |
2, 3 |
289 |
6971852 |
BC067513.1 |
- |
2, 3 |
289 |
6971853 |
BM842783.1 |
- |
2, 3 |
422 |
|
BM851337.1 |
- |
2, 3 |
422 |
|
View Full Product Details