Gene Transcripts

Gene Symbol
ACTR2
Entrez Gene ID
10097
Gene Name
ARP2 actin related protein 2 homolog
Gene Aliases
ARP2
Chromosome Location
Chr. 2: 65227695 - 65271253 on Build GRCh38
UniGene ID
Hs.643727
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005386.2 NP_001005386.1 10 1681
NM_005722.3 NP_005713.1 9 1666

View Full Product Details