Gene Transcripts
- Gene Symbol
- TNFRSF6B
- Entrez Gene ID
- 8771
- Gene Name
- TNF receptor superfamily member 6b
- Gene Aliases
- DCR3, DJ583P15.1.1, M68, M68E, TR6
- Chromosome Location
- Chr. 20: 63696651 - 63698698 on Build GRCh38
- UniGene ID
- Hs.434878
- Species
- Homo sapiens
- Gene Symbol
- RTEL1-TNFRSF6B
- Entrez Gene ID
- 100533107
- Gene Name
- RTEL1-TNFRSF6B readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 20: 63657810 - 63698698 on Build GRCh38
- UniGene ID
- Hs.745057
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037882.1 |
|
38 |
5389 |
|
View Full Product Details