Gene Transcripts

Gene Symbol
LINC01152
Entrez Gene ID
102606463
Gene Name
long intergenic non-protein coding RNA 1152
Gene Aliases
CMPD, TCONS_00025128
Chromosome Location
Chr. 17: 72030654 - 72041297 on Build GRCh38
UniGene ID
Hs.512309
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_110124.1
3 2722
NR_110132.1
3 2212
GenBank mRNA D43770.1 - LIT1584
CMPD
3 2560

View Full Product Details