Gene Transcripts

Gene Symbol
FER
Entrez Gene ID
2241
Gene Name
FER tyrosine kinase
Gene Aliases
PPP1R74, TYK3, p94-Fer
Chromosome Location
Chr. 5: 108746085 - 109196841 on Build GRCh38
UniGene ID
Hs.107418
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308038.1 NP_001294967.1 8 1018
GenBank mRNA BC058030.1 - 8 978

View Full Product Details