Gene Transcripts
- Gene Symbol
- HLA-B
- Entrez Gene ID
- 3106
- Gene Name
- major histocompatibility complex, class I, B
- Gene Aliases
- AS, B-4901, B-5001, Bw-47, Bw-50, HEL-S-83, HLA-B*45ZJ, HLA-B-3506, HLA-B-3905, HLA-B-5502, HLA-B-5602, HLA-B15, HLA-B39, HLA-B49, HLA-B50, HLA-B55, HLA-B59, HLA-B61, HLA-Cw, HLA-DRB1, HLAB, SPDA1
- Chromosome Location
- Chr. 6: 31353866 - 31357245 on Build GRCh38
- UniGene ID
- Hs.654404
- Species
- Homo sapiens
- Gene Symbol
- HLA-H
- Entrez Gene ID
- 3136
- Gene Name
- major histocompatibility complex, class I, H (pseudogene)
- Gene Aliases
- HLAHP
- Chromosome Location
- Chr. 6: 29887760 - 29891080 on Build GRCh38
- UniGene ID
- Hs.728808
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001434.3 |
|
8 |
1348 |
|
GenBank mRNA |
AK090500.1 |
- |
6 |
2142 |
|
- Gene Symbol
- HLA-K
- Entrez Gene ID
- 3138
- Gene Name
- major histocompatibility complex, class I, K (pseudogene)
- Gene Aliases
- HLA-70, HLA70, HLAK
- Chromosome Location
- Chr. 6: 29926659 - 29929839 on Build GRCh38
- UniGene ID
- Hs.728972
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092921.1 |
- |
5 |
2463 |
|
BC035647.1 |
- |
5 |
1612 |
|
View Full Product Details