Gene Transcripts

Gene Symbol
CNTN1
Entrez Gene ID
1272
Gene Name
contactin 1
Gene Aliases
F3, GP135, MYPCN
Chromosome Location
Chr. 12: 40692439 - 41072412 on Build GRCh38
UniGene ID
Hs.143434
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256063.1 NP_001242992.1 13 1653
NM_001256064.1 NP_001242993.1 13 1866
NM_001843.3 NP_001834.2 13 1653
NM_175038.2 NP_778203.1 11 1469
XM_005268651.2 XP_005268708.1 13 2016
XM_006719241.2 XP_006719304.1 13 1640
XM_011537926.2 XP_011536228.1 13 1605
XM_011537927.2 XP_011536229.1 13 1662
XM_017018821.1 XP_016874310.1 13 1656
XM_017018822.1 XP_016874311.1 13 1605
XM_017018823.1 XP_016874312.1 13 2016
XM_017018824.1 XP_016874313.1 13 1640
XM_017018825.1 XP_016874314.1 13 1605
XM_017018826.1 XP_016874315.1 13 1640
XM_017018827.1 XP_016874316.1 13 1662
GenBank mRNA AK289544.1 - 13 1745
BC036569.2 - 13 1592 5273941
Z21488.1 - 13 1547

View Full Product Details